Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGGGTCTTTGCAAAAGTCCACT[C/T]GGCGCTGAGAAACTTTCCTGTTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601532 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP6 - caspase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001226.3 | 590 | Missense Mutation | CAA,CGA | Q,R 260 | NP_001217.2 | |
NM_032992.2 | 590 | Missense Mutation | CAA,CGA | Q,R 171 | NP_116787.1 |
MCUB - mitochondrial calcium uniporter dominant negative beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017918.4 | 590 | Intron | NP_060388.2 | |||
XM_006714246.3 | 590 | Intron | XP_006714309.1 |