Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCGCAGTCCCGCTGGGGGAGGG[C/T]GTCGCGGGCATCGGTCTCACCTGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607827 MIM: 607828 | ||||||||||||||||||||
Literature Links: |
OTOP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OTOP2 - otopetrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOP3 - otopetrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272005.1 | 50 | UTR 5 | NP_001258934.1 | |||
NM_178233.2 | 50 | Missense Mutation | GCG,GTG | A,V 17 | NP_839947.1 | |
XM_011524744.2 | 50 | UTR 5 | XP_011523046.1 |