Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTCTTCCAGACTCGGTGCTGT[A/C]CCCGCCGTGAGTTTCTTGGTTTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C2orf16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf16 - chromosome 2 open reading frame 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032266.3 | 168 | Intron | NP_115642.3 |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271286.1 | 168 | Silent Mutation | GTA,GTC | V,V 8 | NP_001258215.1 | |
NM_001271287.1 | 168 | UTR 5 | NP_001258216.1 | |||
NM_001271288.1 | 168 | UTR 5 | NP_001258217.1 | |||
NM_001271289.1 | 168 | Intron | NP_001258218.1 | |||
NM_001271318.1 | 168 | UTR 5 | NP_001258247.1 | |||
NM_032434.3 | 168 | Silent Mutation | GTA,GTC | V,V 8 | NP_115810.2 |