Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTCGTCATCCTGAGAATTATCA[A/G]TGGGAAAACTGGAGTCTAGAAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2orf69 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2orf69 - chromosome 2 open reading frame 69 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153689.5 | 558 | Silent Mutation | CAA,CAG | Q,Q 125 | NP_710156.3 |
TYW5 - tRNA-yW synthesizing protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |