Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTCCCCACCAATGAAGGATGCA[A/G]CCAAGCAGCAGGTGGGATGAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTBK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTBK1 - tau tubulin kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011514947.2 | Intron | XP_011513249.1 | ||||
XM_011514948.2 | Intron | XP_011513250.1 | ||||
XM_017011364.1 | Intron | XP_016866853.1 | ||||
XM_017011365.1 | Intron | XP_016866854.1 | ||||
XM_017011366.1 | Intron | XP_016866855.1 | ||||
XM_017011367.1 | Intron | XP_016866856.1 | ||||
XM_017011368.1 | Intron | XP_016866857.1 |