Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGCGTTGCGCCCGACTCTCCGCG[A/G]TTACCTGTGCCTAGAGGTGATTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615556 MIM: 603405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAT1 - alpha tubulin acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031722.3 | 553 | Intron | NP_001026892.1 | |||
NM_001190724.2 | 553 | Intron | NP_001177653.1 | |||
NM_001254952.2 | 553 | Intron | NP_001241881.1 | |||
NM_001318762.1 | 553 | Intron | NP_001305691.1 | |||
NM_001318763.1 | 553 | Intron | NP_001305692.1 | |||
NM_024909.3 | 553 | Intron | NP_079185.2 | |||
XM_005249415.2 | 553 | Intron | XP_005249472.1 | |||
XM_011514918.2 | 553 | Intron | XP_011513220.1 | |||
XM_017011316.1 | 553 | Intron | XP_016866805.1 |
C6orf136 - chromosome 6 open reading frame 136 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109938.2 | 553 | Intron | NP_001103408.1 | |||
NM_001161376.1 | 553 | Silent Mutation | CGA,CGG | R,R 120 | NP_001154848.1 | |
NM_145029.3 | 553 | Intron | NP_659466.2 | |||
XM_011514384.1 | 553 | Intron | XP_011512686.1 |
DHX16 - DEAH-box helicase 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |