Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAGGGAACAAGGGGGCTGTGGG[C/T]GTCTCCTTCATGTTTAATGGCACCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600829 MIM: 602753 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INPPL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INPPL1 - inositol polyphosphate phosphatase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001567.3 | 2006 | Silent Mutation | GGC,GGT | G,G 545 | NP_001558.3 | |
XM_005273978.3 | 2006 | Silent Mutation | GGC,GGT | G,G 567 | XP_005274035.1 | |
XM_005273979.4 | 2006 | Silent Mutation | GGC,GGT | G,G 567 | XP_005274036.1 | |
XM_011544999.2 | 2006 | Silent Mutation | GGC,GGT | G,G 545 | XP_011543301.1 | |
XM_011545000.2 | 2006 | Silent Mutation | GGC,GGT | G,G 567 | XP_011543302.1 | |
XM_017017673.1 | 2006 | Silent Mutation | GGC,GGT | G,G 557 | XP_016873162.1 |
PHOX2A - paired like homeobox 2a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |