Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCCACTTACCTGTGTCCTCACC[A/G]CTTTCCTCCTCGCTGGAGGCACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142705 MIM: 603144 MIM: 606936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HRC - histidine rich calcium binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002152.2 | 2005 | Silent Mutation | AGC,AGT | S,S 606 | NP_002143.1 | |
XM_017026733.1 | 2005 | Silent Mutation | AGC,AGT | S,S 606 | XP_016882222.1 |
PPFIA3 - PTPRF interacting protein alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRPM4 - transient receptor potential cation channel subfamily M member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |