Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCTTTTGGGGGCCTCTGAGGC[A/G]TCTGGGCAGGTCCTGCAGAGAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614142 MIM: 610287 MIM: 602327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUEDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUEDC2 - CUE domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024040.2 | 758 | Missense Mutation | CGC,TGC | R,C 203 | NP_076945.2 |
FBXL15 - F-box and leucine rich repeat protein 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024326.3 | 758 | Intron | NP_077302.3 | |||
XM_005270149.2 | 758 | Intron | XP_005270206.1 | |||
XM_005270150.4 | 758 | Intron | XP_005270207.1 | |||
XM_005270151.3 | 758 | Intron | XP_005270208.1 | |||
XM_017016631.1 | 758 | Intron | XP_016872120.1 |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |