Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGACTGCACTCCTGGTTGTCCTC[A/G]TCCTCCTTGCTGTGGCGCTTCAAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602957 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL22 - C-C motif chemokine ligand 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002990.4 | 91 | Missense Mutation | ATC,GTC | I,V 13 | NP_002981.2 | |
XM_017023531.1 | 91 | Missense Mutation | ATC,GTC | I,V 13 | XP_016879020.1 |