Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGAAGCTCTGCTGGGACCAGG[C/T]ACAACTGCCATTAGGTGCCCCAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616776 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP15 - dual specificity phosphatase 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012644.2 | 125 | Intron | NP_001012662.1 | |||
NM_001320478.1 | 125 | Intron | NP_001307407.1 | |||
NM_001320479.1 | 125 | Intron | NP_001307408.1 | |||
NM_080611.4 | 125 | Intron | NP_542178.2 | |||
NM_177991.2 | 125 | Intron | NP_817130.1 | |||
XM_017027654.1 | 125 | Intron | XP_016883143.1 | |||
XM_017027655.1 | 125 | Intron | XP_016883144.1 | |||
XM_017027656.1 | 125 | Intron | XP_016883145.1 | |||
XM_017027657.1 | 125 | Intron | XP_016883146.1 | |||
XM_017027658.1 | 125 | Intron | XP_016883147.1 | |||
XM_017027659.1 | 125 | Intron | XP_016883148.1 | |||
XM_017027660.1 | 125 | Intron | XP_016883149.1 | |||
XM_017027661.1 | 125 | Intron | XP_016883150.1 | |||
XM_017027662.1 | 125 | Intron | XP_016883151.1 |
TTLL9 - tubulin tyrosine ligase like 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008409.2 | 125 | Silent Mutation | GGC,GGT | G,G 12 | NP_001008409.1 | |
XM_005260309.2 | 125 | Intron | XP_005260366.1 | |||
XM_006723723.2 | 125 | Intron | XP_006723786.1 | |||
XM_006723724.2 | 125 | Intron | XP_006723787.1 | |||
XM_011528617.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526919.1 | |
XM_011528618.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526920.1 | |
XM_011528619.1 | 125 | UTR 5 | XP_011526921.1 | |||
XM_011528620.2 | 125 | Intron | XP_011526922.1 | |||
XM_011528621.1 | 125 | Intron | XP_011526923.1 | |||
XM_011528622.1 | 125 | Intron | XP_011526924.1 | |||
XM_011528623.2 | 125 | Intron | XP_011526925.1 | |||
XM_011528624.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526926.1 | |
XM_011528625.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526927.1 | |
XM_011528626.1 | 125 | UTR 5 | XP_011526928.1 | |||
XM_011528627.2 | 125 | Intron | XP_011526929.1 | |||
XM_011528628.1 | 125 | Intron | XP_011526930.1 | |||
XM_011528629.1 | 125 | Intron | XP_011526931.1 | |||
XM_011528630.1 | 125 | Intron | XP_011526932.1 | |||
XM_011528631.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526933.1 | |
XM_011528632.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526934.1 | |
XM_011528633.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526935.1 | |
XM_011528634.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526936.1 | |
XM_011528635.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526937.1 | |
XM_011528637.2 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526939.1 | |
XM_011528638.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526940.1 | |
XM_011528639.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526941.1 | |
XM_011528640.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526942.1 | |
XM_011528641.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526943.1 | |
XM_011528643.2 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526945.1 | |
XM_011528644.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_011526946.1 | |
XM_011528646.1 | 125 | UTR 5 | XP_011526948.1 | |||
XM_017027694.1 | 125 | Intron | XP_016883183.1 | |||
XM_017027695.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883184.1 | |
XM_017027696.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883185.1 | |
XM_017027697.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883186.1 | |
XM_017027698.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883187.1 | |
XM_017027699.1 | 125 | Intron | XP_016883188.1 | |||
XM_017027700.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883189.1 | |
XM_017027701.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883190.1 | |
XM_017027702.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883191.1 | |
XM_017027703.1 | 125 | Silent Mutation | GGC,GGT | G,G 12 | XP_016883192.1 |