Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCGTGCCTTCTTGGCCCTCCTC[C/T]GCTTCTGGGGGGCTGGCTTCTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATG4D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATG4D - autophagy related 4D cysteine peptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281504.1 | 1973 | Intron | NP_001268433.1 | |||
NM_032885.5 | 1973 | Intron | NP_116274.3 | |||
XM_006722927.1 | 1973 | Intron | XP_006722990.1 |
KRI1 - KRI1 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023008.3 | 1973 | Missense Mutation | CAG,CGG | Q,R 655 | NP_075384.3 | |
XM_011528190.2 | 1973 | Missense Mutation | CAG,CGG | Q,R 537 | XP_011526492.1 |
MIR1238 - microRNA 1238 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |