Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCATCCTCCAGAACTCTCCTGA[A/C]GCCAAAATCTTCTGCCTGGTGCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613433 MIM: 612194 MIM: 616292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAUS6 - HAUS augmin like complex subunit 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RRAGA - Ras related GTP binding A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006570.4 | 643 | Missense Mutation | GAA,GAC | E,D 119 | NP_006561.1 |
SAXO1 - stabilizer of axonemal microtubules 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287049.1 | 643 | Intron | NP_001273978.1 | |||
NM_001287050.1 | 643 | Intron | NP_001273979.1 | |||
NM_153707.3 | 643 | Intron | NP_714918.2 | |||
XM_011517744.1 | 643 | Intron | XP_011516046.1 | |||
XM_011517745.1 | 643 | Intron | XP_011516047.1 | |||
XM_011517746.2 | 643 | Intron | XP_011516048.1 | |||
XM_011517747.2 | 643 | Intron | XP_011516049.1 | |||
XM_017014314.1 | 643 | Intron | XP_016869803.1 | |||
XM_017014315.1 | 643 | Intron | XP_016869804.1 |