Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCTTTATAAAATGATTAGATTT[G/A]AGGATCAAGTTAAAATCCACTATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616821 MIM: 610903 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THSD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THSD1 - thrombospondin type 1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS36 - vacuolar protein sorting 36 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282168.1 | 1661 | UTR 3 | NP_001269097.1 | |||
NM_001282169.1 | 1661 | UTR 3 | NP_001269098.1 | |||
NM_016075.3 | 1661 | UTR 3 | NP_057159.2 |