Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGGCCAGCCAGGCAGAAGCTAGG[C/T]GAGTCATCCAGGTGCACCAGCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602863 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WNT9A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WNT9A - Wnt family member 9A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003395.2 | 1456 | Silent Mutation | TCA,TCG | S,S 295 | NP_003386.1 | |
XM_011544271.2 | 1456 | Silent Mutation | TCA,TCG | S,S 225 | XP_011542573.1 |