Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGCGGGCCAAACCCATCCCGG[A/G]TCAGAAAACATCTGAAGCCGACGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604279 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
JMY - junction mediating and regulatory protein, p53 cofactor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152405.4 | 1001 | Missense Mutation | GAT,GGT | D,G 151 | NP_689618.4 | |
XM_005248430.2 | 1001 | Missense Mutation | GAT,GGT | D,G 151 | XP_005248487.1 | |
XM_011543155.2 | 1001 | Missense Mutation | GAT,GGT | D,G 151 | XP_011541457.1 |