Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGTGTTAACATTTTGATAGTGC[A/G]TGGTTCATAGTAGACAGTCAATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605772 MIM: 607843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EBAG9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EBAG9 - estrogen receptor binding site associated, antigen, 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278938.1 | Intron | NP_001265867.1 | ||||
NM_004215.4 | Intron | NP_004206.1 | ||||
NM_198120.2 | Intron | NP_936056.1 | ||||
XM_017013960.1 | Intron | XP_016869449.1 |
PKHD1L1 - polycystic kidney and hepatic disease 1 (autosomal recessive)-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |