Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTAGAAACTGCTGGAGCGAACCC[A/G]TGCCAGGCGAGAGAATCTTCAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616027 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANLN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANLN - anillin actin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284301.2 | 259 | Missense Mutation | CAT,CGT | H,R 13 | NP_001271230.1 | |
NM_001284302.2 | 259 | Missense Mutation | CAT,CGT | H,R 13 | NP_001271231.1 | |
NM_018685.4 | 259 | Missense Mutation | CAT,CGT | H,R 13 | NP_061155.2 | |
XM_006715746.1 | 259 | Missense Mutation | CAT,CGT | H,R 13 | XP_006715809.1 | |
XM_006715747.3 | 259 | Missense Mutation | CAT,CGT | H,R 13 | XP_006715810.1 | |
XM_017012354.1 | 259 | Missense Mutation | CAT,CGT | H,R 13 | XP_016867843.1 | |
XM_017012355.1 | 259 | Missense Mutation | CAT,CGT | H,R 13 | XP_016867844.1 | |
XM_017012356.1 | 259 | Missense Mutation | CAT,CGT | H,R 13 | XP_016867845.1 |
KIAA0895 - KIAA0895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |