Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGAGGAAGATGAAGAAGAAGA[A/G]AAGAGAGAGGACGGGGGTGCAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605398 MIM: 612383 MIM: 614312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCL16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCL16 - C-X-C motif chemokine ligand 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100812.1 | 444 | Intron | NP_001094282.1 | |||
NM_022059.3 | 444 | Intron | NP_071342.2 |
MED11 - mediator complex subunit 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYND15 - zinc finger MYND-type containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136046.2 | 444 | Silent Mutation | GAA,GAG | E,E 126 | NP_001129518.1 | |
NM_001267822.1 | 444 | Silent Mutation | GAA,GAG | E,E 126 | NP_001254751.1 | |
NM_032265.2 | 444 | Silent Mutation | GAA,GAG | E,E 126 | NP_115641.1 | |
XM_017025218.1 | 444 | Silent Mutation | GAA,GAG | E,E 126 | XP_016880707.1 |