Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATCACTAAAAGAAGATTCGCTA[C/T]CTGAACTTTCTTCTACAGTTTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC53 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC53 - coiled-coil domain containing 53 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301107.1 | 722 | Missense Mutation | GAT,GGT | D,G 186 | NP_001288036.1 | |
NM_016053.3 | 722 | Missense Mutation | GAT,GGT | D,G 187 | NP_057137.1 | |
XM_011538423.1 | 722 | Intron | XP_011536725.1 | |||
XM_017019383.1 | 722 | Intron | XP_016874872.1 |