Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGACCTAGACTGAATTTCCGCA[C/G]TATTTGATGAACTATGACACAACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC168 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC168 - coiled-coil domain containing 168 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146197.1 | 20797 | Missense Mutation | ACT,AGT | T,S 6886 | NP_001139669.1 | |
XM_011521106.1 | 20797 | Missense Mutation | ACT,AGT | T,S 6846 | XP_011519408.1 |