Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCAACCAGGTGGCTGTGGGCTG[C/T]GAGTGCCAGGACTGTCTGTGGGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300254 MIM: 300392 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUV39H1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUV39H1 - suppressor of variegation 3-9 homolog 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282166.1 | 842 | Silent Mutation | TGC,TGT | C,C 192 | NP_001269095.1 | |
NM_003173.3 | 842 | Silent Mutation | TGC,TGT | C,C 181 | NP_003164.1 |
WAS - Wiskott-Aldrich syndrome | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |