Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCGGCCCACCGCTGTCAACATGG[C/G]CCGCGCCGCCCGCGACCTGGCTGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615105 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C19orf53 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C19orf53 - chromosome 19 open reading frame 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC130 - coiled-coil domain containing 130 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRI1 - methylthioribose-1-phosphate isomerase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011528355.2 | 387 | Missense Mutation | GCC,GGC | A,G 97 | XP_011526657.1 | |
XM_011528356.2 | 387 | Missense Mutation | GCC,GGC | A,G 97 | XP_011526658.1 | |
XM_011528357.2 | 387 | Missense Mutation | GCC,GGC | A,G 97 | XP_011526659.1 |