Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGGGCCACGGGCCACTCGGGG[G/T]GCGGGTGCATCAGCCAGGGCCGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611683 | ||||||||||||||||||||
Literature Links: |
FN3KRP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FN3KRP - fructosamine 3 kinase related protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024619.3 | 114 | Missense Mutation | GGC,TGC | G,C 22 | NP_078895.2 |
LOC101929552 - uncharacterized LOC101929552 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |