Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACGCCCTGCGCCTGGTGAACCAA[A/G]GTGAGGCGCCGGCCGTTAACTGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610421 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KHDRBS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KHDRBS3 - KH RNA binding domain containing, signal transduction associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006558.2 | 490 | Missense Mutation | AAA,GAA | K,E 30 | NP_006549.1 | |
XM_005250757.2 | 490 | Intron | XP_005250814.1 | |||
XM_005250758.3 | 490 | Missense Mutation | AAA,GAA | K,E 30 | XP_005250815.1 | |
XM_011516796.2 | 490 | Intron | XP_011515098.1 | |||
XM_011516797.2 | 490 | Intron | XP_011515099.1 | |||
XM_011516798.1 | 490 | Missense Mutation | AAA,GAA | K,E 30 | XP_011515100.1 | |
XM_011516799.1 | 490 | Missense Mutation | AAA,GAA | K,E 30 | XP_011515101.1 |