Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGAATTTTATGTAAACATGGA[A/G]ACAACTTTTCTGCCACTAAACGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613567 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC653160 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC653160 - uncharacterized LOC653160 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM35B - transmembrane protein 35B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZMYM6 - zinc finger MYM-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007167.3 | 3296 | Missense Mutation | TCT,TTT | S,F 1023 | NP_009098.3 |