Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGGGAGAAGATCTGACGGAAGAG[A/G]AAGATGGCGGAATCATTCGCAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP4 - FK506 binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002014.3 | 634 | Missense Mutation | AAA,GAA | K,E 146 | NP_002005.1 | |
XM_011520929.2 | 634 | Missense Mutation | AAA,GAA | K,E 101 | XP_011519231.1 |