Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTAATAATCTTGACATGGTTTGAT[T/A]ATCACTTGGTTTATTCTGATTACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETNPPL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ETNPPL - ethanolamine-phosphate phospho-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146590.1 | 1744 | UTR 3 | NP_001140062.1 | |||
NM_001146627.1 | 1744 | UTR 3 | NP_001140099.1 | |||
NM_031279.3 | 1744 | UTR 3 | NP_112569.2 | |||
XM_006714290.2 | 1744 | UTR 3 | XP_006714353.1 |