Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGATTCCAGTGTTACCTCTGGT[C/T]GTAAGACAGCGCCATGGCCCGGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606453 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRBA - LPS responsive beige-like anchor protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199282.2 | 8948 | Missense Mutation | AAC,GAC | N,D 2820 | NP_001186211.2 | |
NM_006726.4 | 8948 | Missense Mutation | AAC,GAC | N,D 2832 | NP_006717.2 | |
XM_005263372.3 | 8948 | Missense Mutation | AAC,GAC | N,D 2837 | XP_005263429.1 | |
XM_005263373.2 | 8948 | Missense Mutation | AAC,GAC | N,D 2837 | XP_005263430.1 | |
XM_005263374.3 | 8948 | Missense Mutation | AAC,GAC | N,D 2826 | XP_005263431.1 | |
XM_005263375.3 | 8948 | Missense Mutation | AAC,GAC | N,D 2821 | XP_005263432.1 | |
XM_011532434.2 | 8948 | Missense Mutation | AAC,GAC | N,D 2832 | XP_011530736.1 | |
XM_017008872.1 | 8948 | Missense Mutation | AAC,GAC | N,D 2826 | XP_016864361.1 | |
XM_017008873.1 | 8948 | Missense Mutation | AAC,GAC | N,D 727 | XP_016864362.1 | |
XM_017008874.1 | 8948 | Missense Mutation | AAC,GAC | N,D 725 | XP_016864363.1 |