Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGCGATGTCAGAGAACGGCTCC[C/T]TCGCCAACTGCTGCGAGGCGGGCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600804 | ||||||||||||||||||||
Literature Links: |
MTNR1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTNR1B - melatonin receptor 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005959.3 | 122 | Missense Mutation | CTC,TTC | L,F 7 | NP_005950.1 | |
XM_011542839.2 | 122 | Missense Mutation | CTC,TTC | L,F 7 | XP_011541141.1 | |
XM_017017777.1 | 122 | Intron | XP_016873266.1 |