Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCAGGTGCCAGGCCCGGGCCACC[A/G]CAGGGCTGCCGGGGCAACATGATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACAN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACAN - aggrecan | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAPLN3 - hyaluronan and proteoglycan link protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307952.1 | 918 | Silent Mutation | TGC,TGT | C,C 295 | NP_001294881.1 | |
NM_178232.3 | 918 | Silent Mutation | TGC,TGT | C,C 233 | NP_839946.1 | |
XM_011521261.1 | 918 | Silent Mutation | TGC,TGT | C,C 277 | XP_011519563.1 | |
XM_017021934.1 | 918 | Intron | XP_016877423.1 | |||
XM_017021935.1 | 918 | Silent Mutation | TGC,TGT | C,C 126 | XP_016877424.1 | |
XM_017021936.1 | 918 | Silent Mutation | TGC,TGT | C,C 126 | XP_016877425.1 |