Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACGGGCAGCATAGTTGTGCTG[C/T]TGCTGATGGATGGAGATGTGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYCL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYCL - v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033081.2 | 1211 | Silent Mutation | CAA,CAG | Q,Q 210 | NP_001028253.1 | |
NM_001033082.2 | 1211 | Silent Mutation | CAA,CAG | Q,Q 240 | NP_001028254.2 | |
NM_005376.4 | 1211 | Intron | NP_005367.2 |