Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACTCTGTAGACATAGCATGTCTT[G/T]TATCCTTCACAAAGCCATTCGCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
METTL11B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
METTL11B - methyltransferase like 11B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136107.1 | 16 | Missense Mutation | TGT,TTT | C,F 28 | NP_001129579.1 | |
XM_011509232.2 | 16 | UTR 5 | XP_011507534.1 | |||
XM_011509233.2 | 16 | Intron | XP_011507535.1 | |||
XM_011509234.2 | 16 | Intron | XP_011507536.1 |
MIR3119-1 - microRNA 3119-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3119-2 - microRNA 3119-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |