Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATCAAACTTGCTGCATAGGGGA[A/T]CTGTAAGACAAAAACAAGCAACACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SKOR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SKOR2 - SKI family transcriptional corepressor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037802.1 | 2986 | Intron | NP_001032891.1 | |||
NM_001278063.1 | 2986 | Missense Mutation | ATC,TTC | I,F 996 | NP_001264992.1 | |
XM_006722525.3 | 2986 | Missense Mutation | ATC,TTC | I,F 996 | XP_006722588.1 |