Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGAATTGGAGGACAGTTTCTTT[C/T]GACTTCGCGAAGATCACATGTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605446 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPGRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPGRIP1 - retinitis pigmentosa GTPase regulator interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020366.3 | 154 | Silent Mutation | CGA,TGA | R,* 52 | NP_065099.3 | |
XM_005267879.2 | 154 | Intron | XP_005267936.1 | |||
XM_005267880.2 | 154 | Intron | XP_005267937.1 | |||
XM_005267881.3 | 154 | Intron | XP_005267938.1 | |||
XM_011536978.1 | 154 | Intron | XP_011535280.1 | |||
XM_011536979.1 | 154 | Intron | XP_011535281.1 | |||
XM_011536980.1 | 154 | Intron | XP_011535282.1 | |||
XM_011536981.1 | 154 | Intron | XP_011535283.1 | |||
XM_011536982.1 | 154 | Intron | XP_011535284.1 | |||
XM_017021473.1 | 154 | Intron | XP_016876962.1 |