Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGTTCCAGTGCACCATCGACGA[C/T]CCCACCAAGCAGACCAAGTTCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX18 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNX18 - sorting nexin 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102575.1 | 1036 | Silent Mutation | GAC,GAT | D,D 282 | NP_001096045.1 | |
NM_001145427.1 | 1036 | Silent Mutation | GAC,GAT | D,D 282 | NP_001138899.1 | |
NM_052870.2 | 1036 | Silent Mutation | GAC,GAT | D,D 282 | NP_443102.2 | |
XM_017008997.1 | 1036 | Silent Mutation | GAC,GAT | D,D 282 | XP_016864486.1 |