Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGCCAGTCCACCACCTACTGCC[A/G]CAAAGGTTAGGACACCAGTGTAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600724 MIM: 610264 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNGB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNGB1 - cyclic nucleotide gated channel beta 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEPP - testis, prostate and placenta expressed | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_199046.2 | 420 | Missense Mutation | CAC,CGC | H,R 128 | NP_950247.2 | |
NM_199456.2 | 420 | Missense Mutation | CAC,CGC | H,R 128 | NP_955535.2 | |
XM_017023218.1 | 420 | Missense Mutation | CAC,CGC | H,R 73 | XP_016878707.1 |