Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGCTGGCTCCAGGGAAGGGCT[A/G]CGTTTGGCCTGGGAGAGTAAGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf132 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf132 - chromosome 14 open reading frame 132 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252507.2 | 281 | Intron | NP_001239436.1 | |||
NM_001282463.1 | 281 | Intron | NP_001269392.1 | |||
NM_001282464.1 | 281 | Intron | NP_001269393.1 | |||
NM_001289139.1 | 281 | Missense Mutation | TAC,TGC | Y,C 35 | NP_001276068.1 | |
XM_017021469.1 | 281 | Intron | XP_016876958.1 |