Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGCGAGTGCCCGCCACAAAGAC[G/T]GTGCATCTGCAGTCACGGGCGCGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602129 MIM: 610675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO9B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO9B - myosin IXB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OCEL1 - occludin/ELL domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USE1 - unconventional SNARE in the ER 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018467.3 | 384 | Silent Mutation | ACG,ACT | T,T 108 | NP_060937.1 | |
XM_017026975.1 | 384 | Silent Mutation | ACG,ACT | T,T 108 | XP_016882464.1 | |
XM_017026976.1 | 384 | Silent Mutation | ACG,ACT | T,T 38 | XP_016882465.1 |