Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACCTTTGACAAACCCAAGTCAGA[C/T]GGGGAACAAAAAACAAAAGAAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R9A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R9A - protein phosphatase 1 regulatory subunit 9A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166160.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | NP_001159632.1 | |
NM_001166161.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | NP_001159633.1 | |
NM_001166162.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | NP_001159634.1 | |
NM_001166163.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | NP_001159635.1 | |
NM_017650.2 | 402 | Silent Mutation | GAC,GAT | D,D 40 | NP_060120.2 | |
XM_011516380.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_011514682.1 | |
XM_011516381.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_011514683.1 | |
XM_011516382.2 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_011514684.1 | |
XM_011516383.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_011514685.1 | |
XM_011516389.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_011514691.1 | |
XM_017012394.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867883.1 | |
XM_017012395.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867884.1 | |
XM_017012396.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867885.1 | |
XM_017012397.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867886.1 | |
XM_017012398.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867887.1 | |
XM_017012399.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867888.1 | |
XM_017012400.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867889.1 | |
XM_017012401.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867890.1 | |
XM_017012402.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867891.1 | |
XM_017012403.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867892.1 | |
XM_017012404.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867893.1 | |
XM_017012405.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867894.1 | |
XM_017012406.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867895.1 | |
XM_017012407.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867896.1 | |
XM_017012408.1 | 402 | Intron | XP_016867897.1 | |||
XM_017012409.1 | 402 | Silent Mutation | GAC,GAT | D,D 40 | XP_016867898.1 | |
XM_017012410.1 | 402 | UTR 5 | XP_016867899.1 | |||
XM_017012411.1 | 402 | Intron | XP_016867900.1 |