Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGCCTCAAACTCATGCTGCTCCT[A/G]GAGGTCATTTCAGGTGAGGATGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102574 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTN3 - actinin alpha 3 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001104.3 | 494 | Silent Mutation | CTA,CTG | L,L 83 | NP_001095.2 | |
NM_001258371.2 | 494 | Silent Mutation | CTA,CTG | L,L 126 | NP_001245300.2 |
ZDHHC24 - zinc finger DHHC-type containing 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |