Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTCGGTTTATCAGAAATACCAA[C/T]GAGAGCGGTGAAGAAGTCACCACCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601267 | |||||||||||||||||||||||
Literature Links: |
CCR2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCR2 - C-C motif chemokine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123041.2 | 240 | Silent Mutation | AAC,AAT | N,N 14 | NP_001116513.2 | |
NM_001123396.1 | 240 | Silent Mutation | AAC,AAT | N,N 14 | NP_001116868.1 | |
XM_011534069.1 | 240 | Silent Mutation | AAC,AAT | N,N 14 | XP_011532371.1 |
LOC102724297 - uncharacterized LOC102724297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |