Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAAGACTAAATGTAACATCTAC[A/G]GCATCATTTCTGCTTAGAGTCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300837 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKK - diacylglycerol kinase kappa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013742.3 | 3495 | Silent Mutation | GCC,GCT | A,A 1145 | NP_001013764.1 | |
XM_017029268.1 | 3495 | Intron | XP_016884757.1 |