Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACCACTGAGCAGAAAGGGAGCTG[A/G]CAGCCACAGAGGCCACACTTTCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DACT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DACT2 - dishevelled binding antagonist of beta catenin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286350.1 | 895 | Intron | NP_001273279.1 | |||
NM_001286351.1 | 895 | Silent Mutation | TGC,TGT | C,C 258 | NP_001273280.1 | |
NM_214462.4 | 895 | Intron | NP_999627.2 | |||
XM_011535507.2 | 895 | Intron | XP_011533809.1 |