Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATATCCTCAACACTCCATTTGTA[A/G]GCAAGTTCCTCCACTGCCTTCTTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BUD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BUD13 - BUD13 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159736.1 | 2441 | Silent Mutation | GCC,GCT | A,A 477 | NP_001153208.1 | |
NM_032725.3 | 2441 | Silent Mutation | GCC,GCT | A,A 611 | NP_116114.1 | |
XM_011543035.2 | 2441 | Silent Mutation | GCC,GCT | A,A 578 | XP_011541337.1 |