Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTTGTCTCTCTCTTTCTGCACCT[C/T]CACGAGGCTGGTCTGGGCCATTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300277 MIM: 300312 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL1RAPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL1RAPL2 - interleukin 1 receptor accessory protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017416.1 | Intron | NP_059112.1 | ||||
XM_011530905.2 | Intron | XP_011529207.1 | ||||
XM_017029403.1 | Intron | XP_016884892.1 |
TEX13A - testis expressed 13A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291277.1 | Intron | NP_001278206.1 | ||||
NM_031274.4 | Intron | NP_112564.1 |