Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCACCCCCAGCCCCCAACGCC[C/T]CTGCCGCTGGGGAGGACACCACTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133450 MIM: 602128 MIM: 602220 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EWSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EWSR1 - EWS RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GAS2L1 - growth arrest specific 2 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278730.1 | 710 | Missense Mutation | CCT,TCT | P,S 178 | NP_001265659.1 | |
NM_006478.4 | 710 | Missense Mutation | CCT,TCT | P,S 178 | NP_006469.2 | |
NM_152236.2 | 710 | Intron | NP_689422.1 | |||
XM_011529824.1 | 710 | Missense Mutation | CCT,TCT | P,S 178 | XP_011528126.1 | |
XM_011529825.1 | 710 | Missense Mutation | CCT,TCT | P,S 178 | XP_011528127.1 | |
XM_017028533.1 | 710 | Missense Mutation | CCT,TCT | P,S 178 | XP_016884022.1 |
RASL10A - RAS like family 10 member A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |