Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCCAGCCATATGTGCTTGTGAC[A/T]ATACAGCATGGAGATGTCACCAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 140571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSP90AA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSP90AA1 - heat shock protein 90 alpha family class A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017963.2 | 1987 | Silent Mutation | ATA,ATT | I,I 721 | NP_001017963.2 | |
NM_005348.3 | 1987 | Silent Mutation | ATA,ATT | I,I 599 | NP_005339.3 | |
XM_011536718.2 | 1987 | Silent Mutation | ATA,ATT | I,I 720 | XP_011535020.1 |