Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAATATGCACATGATAAGGCCCA[A/C]GTCCGCCAAGGGACGGACAAGACCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBXN10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBXN10 - UBX domain protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152376.4 | 210 | Missense Mutation | AAG,ACG | K,T 40 | NP_689589.1 | |
XM_005245742.3 | 210 | Missense Mutation | AAG,ACG | K,T 40 | XP_005245799.1 | |
XM_011540699.2 | 210 | Missense Mutation | AAG,ACG | K,T 40 | XP_011539001.1 |
UBXN10-AS1 - UBXN10 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |