Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTCCCTTGTAAAATCCACCGAC[A/G]CCTTCTTTCCTTTAGAGGGAAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610815 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A32 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A32 - solute carrier family 25 member 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030780.4 | 1220 | Silent Mutation | GGC,GGT | G,G 274 | NP_110407.2 | |
XM_017013877.1 | 1220 | Silent Mutation | GGC,GGT | G,G 142 | XP_016869366.1 |